Få mere at vide
2-(2-Pyridinylamino)-benzenethiol Hydrochloride, TRC
Artikelnummer. 30135014
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135014 10 g
30011294 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135014 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99335010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 163275-26-1
Molekylær formel C11H10N2S • HCl
Molekylvægt (g/mol) 202.283646
Synonym Benzenethiol, 2-(2-pyridinylamino)-, hydrochloride (1:1) (ACI), Benzenethiol, 2-(2-pyridinylamino)-, monohydrochloride (9CI), 2-(2-Pyridinylamino)benzenethiol hydrochloride, 2-[(Pyridin-2-yl)amino]benzene-1-thiol hydrochloride
IUPAC navn 2-(pyridin-2-ylamino)benzenethiol;hydrochloride
SMIL SC1=CC=CC=C1NC2=NC=CC=C2.Cl
Tekniske data
Kemisk navn eller materiale 2-(2-Pyridinylamino)-benzenethiol Hydrochloride
Mængde 10 g
InChI formel InChI=1S/C11H10N2S.ClH/c14-10-6-2-1-5-9(10)13-11-7-3-4-8-12-11;/h1-8,14H,(H,12,13);1H
Formel vægt 238.0331