Learn More
2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol, 98%, BLD Pharm™
Product Code. 31029925
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
1kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31029925 25 g -
31018732 1 kg 1kg
31018466 100 g -
31024638 500 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31029925 Supplier BLD Pharm Supplier No. BD11732125g
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 112-60-7
Molecular Formula C8H18O5
MDL Number MFCD00002879
Quantity 25 g
SMILES OCCOCCOCCOCCO
Molecular Weight (g/mol) 194.23
Percent Purity 98%
Chemical Name or Material 2,2'-((Oxybis(ethane-2,1-diyl))bis(oxy))diethanol