missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2-Methoxyphenyl)thiazolidine, 97%

Brand:  Alfa Aesar™ H62646.06

Additional Details : CAS Number : 40790-78-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15469148

  • 8007.80 DKK / 5g


Estimated Shipment date
from Supplier 17-10-2018
Add to basket

Specifications

Chemical Identifiers

40790-78-1
195.28
HJPJOKSJGDFDBQ-UHFFFAOYSA-N
218556
COC1=CC=CC=C1C2NCCS2
C10H13NOS
MFCD00805017
2-(2-Thiazolidinyl)anisole
2-(2-methoxyphenyl)-1,3-thiazolidine

Specifications

2-(2-Methoxyphenyl)thiazolidine
97%
MFCD00805017
5g
COC1=CC=CC=C1C2NCCS2
195.28
195.28
40790-78-1
C10H13NOS
2-(2-Thiazolidinyl)anisole
HJPJOKSJGDFDBQ-UHFFFAOYSA-N
2-(2-methoxyphenyl)-1,3-thiazolidine
218556
Liquid
Videos
SDS