Få mere at vide
2-(2-Methoxyphenoxy)propane-1,3-diol (B-Isomer), Mikromol™
Artikelnummer. 30049539
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049539 25 mg
30049251 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049539 Leverandør Mikromol Leverandørnr. MM0354.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(2-Methoxyphenoxy)propane-1,3-diol (B-Isomer)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(2-Methoxyphenoxy)propane-1,3-diol
CAS 14007-09-1
Analyt- eller komponentnavne 2-(2-Methoxyphenoxy)propane-1,3-diol
Forsendelsestilstand Room Temperature
Molekylær formel C10 H14 O4
Certificeringer/compliance ISO 17025
Synonym Guaifenesin Imp. B (EP), 2-(2-Methoxyphenoxy)propane-1,3-diol, B-Isomer of Guaifenesin, Guaifenesin Impurity B
InChI formel InChI=1S/C10H14O4/c1-13-9-4-2-3-5-10(9)14-8(6-11)7-12/h2-5,8,11-12H,6-7H2,1H3
SMIL COc1ccccc1OC(CO)CO
IUPAC navn 2-(2-methoxyphenoxy)propane-1,3-diol
Molekylvægt (g/mol) 198.22
Mængde 25 mg
Formel vægt 198.0892 g/mol