Få mere at vide
2-(2-Methoxyphenoxy)-1,3-propanediol, TRC
Artikelnummer. 30007296
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007296 500 mg
30004165 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30007296 Leverandør Toronto Research Chemicals Leverandørnr. TRCM271630500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14007-09-1
Molekylær formel C10 H14 O4
Molekylvægt (g/mol) 198.22
Synonym 2-(2-Methoxyphenoxy)propane-1,3-diol, Guaifenesin Imp. B (EP), B-Isomer of Guaifenesin, Isoguaifenesin
IUPAC navn 2-(2-methoxyphenoxy)propane-1,3-diol
SMIL COc1ccccc1OC(CO)CO
Tekniske data
Kemisk navn eller materiale 2-(2-Methoxyphenoxy)propane-1,3-diol
Renhedsgrad noter HPLC
Mængde 500 mg
InChI formel InChI=1S/C10H14O4/c1-13-9-4-2-3-5-10(9)14-8(6-11)7-12/h2-5,8,11-12H,6-7H2,1H3
Formel vægt 198.0892
Procent renhed >95%