Få mere at vide
2-(2-Iodophenyl-d4)acetic Acid, TRC
Artikelnummer. 30398760
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30398760 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30398760 Leverandør Toronto Research Chemicals Leverandørnr. TRCI71753210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(2-Iodophenyl-d4)acetic Acid
CAS 1217360-62-7
Molekylær formel C8H3D4IO2
Mængde 10 mg
Synonym (o-Iodophenyl-d4)acetic Acid, (2-Iodophenyl-d4)acetic Acid, 2-Iodobenzene-d4-acetic Acid
InChI formel InChI=1S/C8H7IO2/c9-7-4-2-1-3-6(7)5-8(10)11/h1-4H,5H2,(H,10,11)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c(CC(=O)O)c(I)c1[2H]
IUPAC navn 2-(2,3,4,5-tetradeuterio-6-iodophenyl)acetic acid
Molekylvægt (g/mol) 266.07
Formel vægt 265.9742