missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxyphenyl)benzothiazole, 98%, Thermo Scientific Chemicals

Catalog Number 11493947
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
3411-95-8
C13H9NOS
227.28
MFCD00022869
NSDGEQWRYXOZLN-UKTHLTGXSA-N
phenol, o-2-benzothiazolyl, 2-2-hydroxyphenyl benzothiazole, o-2-benzothiazolyl phenol, phenol, 2-2-benzothiazolyl, 2-2-benzothiazolyl phenol, 2-o-hydroxyphenyl benzothiazole, 2-2'-hydroxyphenyl benzothiazole, 2-1,3-benzothiazol-2-yl phenol, 6z-6-3h-1,3-benzothiazol-2-ylidene cyclohexa-2,4-dien-1-one, mvvgspcxhrfddr-uhfffaoysa-n
5376551
O=C1C=CC=C\C1=C1\NC2=CC=CC=C2S1
This item is not returnable. View return policy
3411-95-8
C13H9NOS
227.28
MFCD00022869
NSDGEQWRYXOZLN-UKTHLTGXSA-N
phenol, o-2-benzothiazolyl, 2-2-hydroxyphenyl benzothiazole, o-2-benzothiazolyl phenol, phenol, 2-2-benzothiazolyl, 2-2-benzothiazolyl phenol, 2-o-hydroxyphenyl benzothiazole, 2-2'-hydroxyphenyl benzothiazole, 2-1,3-benzothiazol-2-yl phenol, 6z-6-3h-1,3-benzothiazol-2-ylidene cyclohexa-2,4-dien-1-one, mvvgspcxhrfddr-uhfffaoysa-n
5376551
O=C1C=CC=C\C1=C1\NC2=CC=CC=C2S1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 130°C to 134°C
Quantity 5 g
Beilstein 173026
IUPAC Name (6Z)-6-(3H-1,3-benzothiazol-2-ylidene)cyclohexa-2,4-dien-1-one
Formula Weight 227.29
Percent Purity 98%
Chemical Name or Material 2-(2-Hydroxyphenyl)benzothiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'