missing translation for 'onlineSavingsMsg'
Learn More

2-[(2-Hydroxyethyl)octadecylamino]ethyl ester octadecanoic acid, TRC

Product Code. 30395006
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30395006 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395006 Supplier Toronto Research Chemicals Supplier No. TRCH998250100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-[(2-Hydroxyethyl)octadecylamino]ethyl ester octadecanoic acid
CAS 52497-24-2
Molekylær formel C40 H81 N O3
Mængde 100 mg
Synonym Octadecyl(2-hydroxyethyl)(2-stearoyloxyethyl)amine, Octadecyldiethanolamine monostearate, Stearyldiethanolamine monostearate, Stearyldiethanolamine stearate
InChI formel InChI=1S/C40H81NO3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-41(36-38-42)37-39-44-40(43)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h42H,3-39H2,1-2H3
SMIL O=C(OCCN(CCO)CCCCCCCCCCCCCCCCCC)CCCCCCCCCCCCCCCCC
IUPAC navn 2-[2-hydroxyethyl(octadecyl)amino]ethyl octadecanoate
Molekylvægt (g/mol) 624.076
Formel vægt 623.622
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.