missing translation for 'onlineSavingsMsg'
Learn More

2-[(2-Hydroxyethyl)amino]-1-phenylethanol, TRC

Product Code. 30406589
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30406589 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406589 Supplier Toronto Research Chemicals Supplier No. TRCH949493100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-[(2-Hydroxyethyl)amino]-1-phenylethanol
CAS 4397-15-3
Molekylær formel C10H15NO2
Mængde 100 mg
InChI formel InChI=1S/C10H15NO2/c12-7-6-11-8-10(13)9-4-2-1-3-5-9/h1-5,10-13H,6-8H2
SMIL OCCNCC(O)c1ccccc1
IUPAC navn 2-(2-hydroxyethylamino)-1-phenylethanol
Molekylvægt (g/mol) 181.23
Formel vægt 181.1103
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.