Få mere at vide
2-(2-Ethoxyphenyl)-5,7-dimethylimidazo[5,1-f][1,2,4]triazin-4(1H)-one, TRC
Artikelnummer. 30371208
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30371208 10 mg
30374182 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30371208 Leverandør Toronto Research Chemicals Leverandørnr. TRCE67958510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 224789-20-2
Molekylær formel C15 H22 N4 O3
Molekylvægt (g/mol) 306.36
IUPAC navn 3-methyl-7-(5-oxohexyl)-1-propylpurine-2,6-dione
SMIL CCCN1C(=O)N(C)c2ncn(CCCCC(=O)C)c2C1=O
Tekniske data
Kemisk navn eller materiale 2-(2-Ethoxyphenyl)-5,7-dimethylimidazo[5,1-f][1,2,4]triazin-4(1H)-one
Mængde 10 mg
InChI formel InChI=1S/C15H22N4O3/c1-4-8-19-14(21)12-13(17(3)15(19)22)16-10-18(12)9-6-5-7-11(2)20/h10H,4-9H2,1-3H3
Formel vægt 306.169