missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyphenoxy)ethyl bromide, 98%, Thermo Scientific™

Catalog Number p-7048133
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
3259-03-8
MFCD02030483
This item is not returnable. View return policy
3259-03-8
MFCD02030483

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 41°C to 43°C
Molecular Formula C10H13BrO2
Quantity 5 g
Beilstein 6290077
Synonym 2-2-ethoxyphenoxy ethyl bromide, 1-2-bromoethoxy-2-ethoxybenzene, 2-2-ethoxyphenoxy ethylbromide, 2-2-bromoethoxy phenetole, benzene, 1-2-bromoethoxy-2-ethoxy, 1-2-bromoethoxy-2-ethyoxybenzene, 1-2-bromoethoxy-2-ethoxy-benzene, 2-bromo-1-2-ethoxyphenoxy ethane, tamsulosin impurity i, pubchem13149
InChI Key IOYHGBZPUZBUTJ-UHFFFAOYSA-N
SMILES CCOC1=CC=CC=C1OCCBr
IUPAC Name 1-(2-bromoethoxy)-2-ethoxybenzene
Molecular Weight (g/mol) 245.12
PubChem CID 7010183
Formula Weight 245.12
Percent Purity 98%
Chemical Name or Material 2-(2-Ethoxyphenoxy)ethyl bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'