Få mere at vide
2,2’-(Ethanediyldiimino)bis-butanoic Acid(Mixture of Diastereomers), TRC
Artikelnummer. 30376403
Skift visning
Mængde:
100 mg
500 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376403 500 mg
30375624 100 mg
30376760 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30376403 Leverandør Toronto Research Chemicals Leverandørnr. TRCE890340500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 498-17-9
Molekylær formel C10 H20 N2 O4
Molekylvægt (g/mol) 232.28
Synonym 2,2'-(Ethylenediimino)di-butyric Acid, Ethylenediamine-N,N'-di-α-butyric Acid
IUPAC navn 2-[2-(1-carboxypropylamino)ethylamino]butanoic acid
SMIL CCC(NCCNC(CC)C(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale 2,2'-(Ethanediyldiimino)bis-butanoic acid
Mængde 500 mg
InChI formel InChI=1S/C10H20N2O4/c1-3-7(9(13)14)11-5-6-12-8(4-2)10(15)16/h7-8,11-12H,3-6H2,1-2H3,(H,13,14)(H,15,16)
Formel vægt 232.1423