Få mere at vide
2,2-Diphenylhydantoic Acid, TRC
Artikelnummer. 30379804
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30379804 50 mg
30376288 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30379804 Leverandør Toronto Research Chemicals Leverandørnr. TRCD49166050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6802-95-5
Molekylær formel C15 H14 N2 O3
Molekylvægt (g/mol) 270.28
Synonym (Carbamoylamino)-(diphenyl)acetic Acid, Phenytoin Imp. E (EP), Phenytoin Sodium Imp. E (EP)
IUPAC navn 2-(carbamoylamino)-2,2-diphenylacetic acid
SMIL NC(=O)NC(C(=O)O)(c1ccccc1)c2ccccc2
Tekniske data
Kemisk navn eller materiale (Carbamoylamino)(diphenyl)acetic Acid
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C15H14N2O3/c16-14(20)17-15(13(18)19,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,(H,18,19)(H3,16,17,20)
Formel vægt 270.1004
Procent renhed >95%