Få mere at vide
2,2-dimethylhexanal, TRC
Artikelnummer. 30360994
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30360994 500 mg
30351925 1 g
30349061 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30360994 Leverandør Toronto Research Chemicals Leverandørnr. TRCD206716500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 996-12-3
Molekylær formel C8H16O
Molekylvægt (g/mol) 128.21
IUPAC navn 2,2-dimethylhexanal
SMIL CCCCC(C)(C)C=O
Tekniske data
Kemisk navn eller materiale 2,2-dimethylhexanal
Mængde 500 mg
InChI formel InChI=1S/C8H16O/c1-4-5-6-8(2,3)7-9/h7H,4-6H2,1-3H3
Formel vægt 128.1201