Få mere at vide
2,2-Dimethylbutyric-d11 Acid, CDN
Artikelnummer. 30316755
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30316755 1 g
30310706 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316755 Leverandør CDN Leverandørnr. CDND58431G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,2-Dimethylbutanoic Acid-D11
CAS 1219804-04-2
Molekylær formel C6 D11 H O2
Mængde 1 g
Synonym 2,2-Dimethylbutanoic-d11 Acid, 2,2-Di(methyl-d3)butanoic-3,3,4,4,4-d5 Acid, 2,2-Dimethylbutanoic Acid-D11, 2,2-Dimethylbutyric-d11 acid, tert-Amylcarboxylic acid-d11, α,α-Dimethylbutyric-d11 acid
InChI formel InChI=1 S/C6H12O2/c1-4-6(2,3)5(7)8/h4H2,1-3H3,(H,7,8)/i1D3,2D3,3D3,4D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C(C(=O)O)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 3,3,4,4,4-pentadeuterio-2,2-bis(trideuteriomethyl)butanoic acid
Molekylvægt (g/mol) 127.2261
Formel vægt 127.1528 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity