missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,2-Dimethyl-3-butenoic acid, 95%

Brand:  Alfa Aesar™ H64067.MD

Additional Details : CAS Number : 10276-09-2 Weight : 0.00025kg Transport : UN number : 3265 Chem class : 8 Pack group : III

Quantity 250mg

 View more versions of this product

Product Code. 15481099

  • 367.68 DKK / 250mg


In stock, ready to ship: 1
Add to basket

Specifications

Chemical Identifiers

10276-09-2
114.144
SCFWAOWWAANBPY-UHFFFAOYSA-N
139122
CC(C)(C=C)C(=O)O
C6H10O2
MFCD00196967
3-butenoic acid, 2,2-dimethyl, 2,2-dimethyl-but-3-enoic acid, 2,2-dimethyl-3-butenoic acid, ch2=chc ch3 2cooh, 3-butenoic acid,2,2-dimethyl
2,2-dimethylbut-3-enoic acid

Specifications

2,2-Dimethyl-3-butenoic acid
1.4295
Colorless
250mg
C6H10O2
UN3265
3-butenoic acid, 2,2-dimethyl, 2,2-dimethyl-but-3-enoic acid, 2,2-dimethyl-3-butenoic acid, ch2=chc ch3 2cooh, 3-butenoic acid,2,2-dimethyl
CC(C)(C=C)C(=O)O
114.144
114.14
95%
0.957 g/mL
185°C
-6°C
10276-09-2
MFCD00196967
1700758
SCFWAOWWAANBPY-UHFFFAOYSA-N
2,2-dimethylbut-3-enoic acid
139122
Liquid
Videos
SDS Documents