Få mere at vide
2,2-Difluoromalonamide, TRC
Artikelnummer. 30367444
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367444 100 mg
30350906 10 mg
30358110 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30367444 Leverandør Toronto Research Chemicals Leverandørnr. TRCD357845100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 425-99-0
Molekylær formel C3H4N2O2F2
Molekylvægt (g/mol) 138.07
IUPAC navn 2,2-difluoropropanediamide
SMIL NC(=O)C(F)(F)C(=O)N
Tekniske data
Kemisk navn eller materiale 2,2-Difluoromalonamide
Mængde 100 mg
InChI formel InChI=1S/C3H4F2N2O2/c4-3(5,1(6)8)2(7)9/h(H2,6,8)(H2,7,9)
Formel vægt 138.0241