Få mere at vide
2,2-Difluoro-2-(thiophen-2-yl)ethan-1-amine, TRC
Artikelnummer. 30366777
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366777 100 mg
30350930 500 mg
30366677 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30366777 Leverandør Toronto Research Chemicals Leverandørnr. TRCD206081100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1555527-48-4
Molekylær formel C6H7F2NS
Molekylvægt (g/mol) 163.19
Synonym beta,beta-Difluoro-2-thiopheneethanamine
IUPAC navn 2,2-difluoro-2-thiophen-2-ylethanamine
SMIL NCC(F)(F)c1cccs1
Tekniske data
Kemisk navn eller materiale 2,2-Difluoro-2-(thiophen-2-yl)ethan-1-amine
Mængde 100 mg
InChI formel InChI=1S/C6H7F2NS/c7-6(8,4-9)5-2-1-3-10-5/h1-3H,4,9H2
Formel vægt 163.0267