Learn More
2-(2-Cyanophenyl)benzimidazole, 97%, Thermo Scientific™
Catalog Number 15484498
Click to view available options
Quantity:
1 g
5 g
76889-76-4
C14H9N3
219.247
MFCD11218394
QDFTUAOALSHOGT-UHFFFAOYSA-N
2-1h-benzo d imidazol-2-yl benzonitrile, 2-2-cyanophenyl benzimidazole, 2-1h-1,3-benzodiazol-2-yl benzonitrile, 2-cyanophenylbenzimidazole, 2-2'-cyanophenyl benzimidazole, 2-1h-benzoimidazole-2-yl benzonitrile
12608868
2-(1H-benzimidazol-2-yl)benzonitrile
C1=CC=C(C(=C1)C#N)C2=NC3=CC=CC=C3N2
76889-76-4
C14H9N3
219.247
MFCD11218394
QDFTUAOALSHOGT-UHFFFAOYSA-N
2-1h-benzo d imidazol-2-yl benzonitrile, 2-2-cyanophenyl benzimidazole, 2-1h-1,3-benzodiazol-2-yl benzonitrile, 2-cyanophenylbenzimidazole, 2-2'-cyanophenyl benzimidazole, 2-1h-benzoimidazole-2-yl benzonitrile
12608868
2-(1H-benzimidazol-2-yl)benzonitrile
C1=CC=C(C(=C1)C#N)C2=NC3=CC=CC=C3N2
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3439
Beilstein 5947339
Formula Weight 219.25
Percent Purity 97%
Chemical Name or Material 2-(2-Cyanophenyl)benzimidazole