missing translation for 'onlineSavingsMsg'
Få mere at vide

2-[(2-Chlorophenyl)amino]-3-nitrobenzoic Acid, TRC

Artikelnummer. 30368552
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30368552 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368552 Supplier Toronto Research Chemicals Supplier No. TRCC37762810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-[(2-Chlorophenyl)amino]-3-nitrobenzoic Acid
CAS 38120-54-6
Molekylær formel C13H9ClN2O4
Mængde 10 mg
Synonym N-(2-Chlorophenyl)-3-nitroanthranilic Acid, 2'-Chloro-6-nitrodiphenylamine-2-carboxylic Acid
InChI formel InChI=1S/C13H9ClN2O4/c14-9-5-1-2-6-10(9)15-12-8(13(17)18)4-3-7-11(12)16(19)20/h1-7,15H,(H,17,18)
SMIL O=C(C1=CC=CC(N(=O)=O)=C1NC2=C(Cl)C=CC=C2)O
IUPAC navn 2-((2-chlorophenyl)amino)-3-nitrobenzoic acid
Molekylvægt (g/mol) 292.67
Formel vægt 292.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.