Learn More
2-(2-Chlorophenyl)-2,3-dihydrobenzo[d]oxazole-5-carboxylic Acid, TRC
Product Code. 30354910
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30354910 50 mg
30360175 100 mg
30353427 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30354910 Supplier Toronto Research Chemicals Supplier No. TRCC72301550MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1018498-30-0
Molecular Formula C14 H10 Cl N O3
Molecular Weight (g/mol) 275.687
Synonym 2-(2-Chlorophenyl)-1,3-Benzoxazole-5-Carboxylic Acid
IUPAC Name 2-(2-chlorophenyl)-2,3-dihydro-1,3-benzoxazole-5-carboxylic acid
SMILES OC(=O)c1ccc2OC(Nc2c1)c3ccccc3Cl
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C14H10ClNO3/c15-10-4-2-1-3-9(10)13-16-11-7-8(14(17)18)5-6-12(11)19-13/h1-7,13,16H,(H,17,18)
Formula Weight 275.035
Chemical Name or Material 2-(2-Chlorophenyl)-2,3-dihydrobenzo[d]oxazole-5-carboxylic Acid