Få mere at vide
2-(2-Chloroethyl-d4)-1H-isoindole-1,3-dione, TRC
Artikelnummer. 30361385
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30361385 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361385 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36610210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(2-Chloroethyl-d4)-1H-isoindole-1,3-dione
CAS 1252995-10-0
Molekylær formel C10H4D4ClNO2
Mængde 10 mg
Synonym 2-(2-Chloroethyl-1,1,2,2-d4)-1H-isoindole-1,3(2H)-dione
InChI formel InChI=1S/C10H8ClNO2/c11-5-6-12-9(13)7-3-1-2-4-8(7)10(12)14/h1-4H,5-6H2/i5D2,6D2
SMIL [2H]C([2H])(Cl)C([2H])([2H])N1C(=O)c2ccccc2C1=O
IUPAC navn 2-(2-chloro-1,1,2,2-tetradeuterioethyl)isoindole-1,3-dione
Molekylvægt (g/mol) 213.65
Formel vægt 213.0495