Få mere at vide
2-(2-Chloroethoxy)-N-(4-nitrophenyl)acetamide-d4, TRC
Artikelnummer. 30365184
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30365184 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365184 Leverandør Toronto Research Chemicals Leverandørnr. TRCC365727250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(2-Chloroethoxy)-N-(4-nitrophenyl)acetamide-d4
CAS 1329612-84-1
Molekylær formel C10H7D4ClN2O4
Mængde 250 mg
InChI formel InChI=1S/C10H11ClN2O4/c11-5-6-17-7-10(14)12-8-1-3-9(4-2-8)13(15)16/h1-4H,5-7H2,(H,12,14)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1NC(=O)COCCCl)[N+](=O)[O-]
IUPAC navn 2-(2-chloroethoxy)-N-(2,3,5,6-tetradeuterio-4-nitrophenyl)acetamide
Molekylvægt (g/mol) 262.68
Formel vægt 262.0658