missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chloroethoxy)ethylphosphonic Acid, TRC

Product Code. 30341911
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30341911 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341911 Supplier Toronto Research Chemicals Supplier No. TRCB406220500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-(2-Chloroethoxy)ethylphosphonic Acid
CAS 69404-51-9
Molekylær formel C4H10O4PCl
Mængde 500 mg
Synonym [2-(2-Chloroethoxy)ethyl]phosphonic Acid
InChI formel InChI=1S/C4H10ClO4P/c5-1-2-9-3-4-10(6,7)8/h1-4H2,(H2,6,7,8)
SMIL OP(=O)(O)CCOCCCl
IUPAC navn 2-(2-chloroethoxy)ethylphosphonic acid
Molekylvægt (g/mol) 188.55
Formel vægt 188.0005
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.