missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Bromophenyl)benzimidazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-7039197
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1053659-26-9
C14H9BrN2O2
317.14
MFCD11656167
VDQXOCQBIBFAIF-UHFFFAOYSA-N
2-2-bromophenyl-1h-benzoimidazole-5-carboxylic acid, 2-2-bromophenyl-1h-1,3-benzodiazole-5-carboxylic acid, 2-2-bromophenyl benzimidazole-5-carboxylic acid, 2-2-bromophenyl-1h-benzimidazole-6-carboxylic acid, 2-2-bromophenyl-1h-benzoimidazole-5-carboxylic acid, technical grade
43349242
OC(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1Br
This item is not returnable. View return policy
1053659-26-9
C14H9BrN2O2
317.14
MFCD11656167
VDQXOCQBIBFAIF-UHFFFAOYSA-N
2-2-bromophenyl-1h-benzoimidazole-5-carboxylic acid, 2-2-bromophenyl-1h-1,3-benzodiazole-5-carboxylic acid, 2-2-bromophenyl benzimidazole-5-carboxylic acid, 2-2-bromophenyl-1h-benzimidazole-6-carboxylic acid, 2-2-bromophenyl-1h-benzoimidazole-5-carboxylic acid, technical grade
43349242
OC(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 23490987
IUPAC Name 2-(2-bromophenyl)-3H-benzimidazole-5-carboxylic acid
Formula Weight 317.14
Percent Purity 97%
Chemical Name or Material 2-(2-Bromophenyl)benzimidazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'