missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bithiophene-5-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 210.28 |
Percent Purity | 97% |
Chemical Name or Material | 2, 2'-Bithiophene-5-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'