missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bis(diphenylphosphino)biphenyl, 98%, Thermo Scientific Chemicals

Catalog Number p-7030655
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
84783-64-2
C36H28P2
522.57
MFCD03094574
GRTJBNJOHNTQBO-UHFFFAOYSA-N
2,2'-bis diphenylphosphino-1,1'-biphenyl, 2,2'-bis diphenylphosphino biphenyl, biphep, 2-2-diphenylphosphanylphenyl phenyl-diphenyl-phosphane, dpbp, 2'-diphenylphosphanyl-1,1'-biphenyl-2-yl diphenylphosphane, pubchem9102, acmc-209pwq, 2,2'-bis diphenylphosphino-biphenyl, ar-2,2'-bis diphenylphosphino biphenyl
2734940
[2-(2-diphenylphosphanylphenyl)phenyl]-diphenylphosphane
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
84783-64-2
C36H28P2
522.57
MFCD03094574
GRTJBNJOHNTQBO-UHFFFAOYSA-N
2,2'-bis diphenylphosphino-1,1'-biphenyl, 2,2'-bis diphenylphosphino biphenyl, biphep, 2-2-diphenylphosphanylphenyl phenyl-diphenyl-phosphane, dpbp, 2'-diphenylphosphanyl-1,1'-biphenyl-2-yl diphenylphosphane, pubchem9102, acmc-209pwq, 2,2'-bis diphenylphosphino-biphenyl, ar-2,2'-bis diphenylphosphino biphenyl
2734940
[2-(2-diphenylphosphanylphenyl)phenyl]-diphenylphosphane
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 211°C to 215°C
Quantity 250 mg
Formula Weight 522.57
Percent Purity 98%
Chemical Name or Material 2,2'-Bis(diphenylphosphino)biphenyl

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'