missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene]chloro(p-cymene)ruthenium Chloride, TRC
Specifications
Specifications
| Kemisk navn eller materiale | [2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene]chloro(p-cymene)ruthenium Chloride |
| CAS | 130004-33-0 |
| Molekylær formel | C45 H35 Cl P2 Ru . C10 H20 . Cl |
| Mængde | 250 mg |
| Synonym | [1,1'-(1S)-[1,1'-Binaphthalene]-2,2'-diylbis[1,1-diphenylphosphine-κP]]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]ruthenium(1+) Chloride, (S)-[[1,1'-Binaphthalene]-2,2'-diylbis[diphenylphosphine]-P,P']chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]ruthenium(1+) Chloride, (S)-[1,1'-Binaphthalene]-2,2'-diylbis[diphenylphosphine Ruthenium Complex, ((S)-(2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl))(chloro)(η6-p-isopropyltoluene)ruthenium(1+) Chloride, ((S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl)(chloro)(η6-4-isopropyltoluene)ruthenium(1+) Chloride, [(1S)-[1,1'-Binaphthalene]-2,2'-diylbis[diphenylphosphine-κP]]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]ruthenium(1+) Chloride |
| InChI formel | InChI=1S/C44H32P2.C10H20.CH3.2ClH.Ru/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38;1-8(2)10-6-4-9(3)5-7-10;;;;/h1-32H;8-10H,4-7H2,1-3H3;1H3;2*1H;/q;;;;;+2/p-2/t;9-,10+;;;; |
| SMIL | [Cl-].[Cl-][Ru+2]1(C)[P](c2ccccc2)(c3ccccc3)c4ccc5ccccc5c4c6c(ccc7ccccc67)[P]1(c8ccccc8)c9ccccc9.CC(C)[C@@H]%10CC[C@H](C)CC%10 |
| IUPAC navn | dichlororuthenium;[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;1-methyl-4-propan-2-ylbenzene |
| Molekylvægt (g/mol) | 949.949 |
| Formel vægt | 949.22 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?