Få mere at vide
2,2-Bis(4-hydroxyphenyl)-1,1,1-trichloroethane-d8 (Major), TRC
Artikelnummer. 30331884
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30331884 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331884 Leverandør Toronto Research Chemicals Leverandørnr. TRCB4474025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,2-Bis(4-hydroxyphenyl)-1,1,1-trichloroethane-d8
CAS 1794780-53-2
Molekylær formel C14H3D8Cl3O2
Mængde 5 mg
Synonym 4,4'-(2,2,2-Trichloroethylidene)diphenol-d8, 4,4'-Dihydroxydiphenyltrichloroethane-d8, 2,2-Bis(4-hydroxyphenyl)-1,1,1-trichloroethane-d8, Bisdemethylmethoxychlor-d8, HPTE-d8, 1,1,1-Trichloro-2,2-bis(4-hydroxyphenyl)ethane-d8, Hydroxychlor-d8, NSC 7045-d8, p,p'-Hydroxy-DDT-d8, 1,1-Bis(4-hydroxyphenyl)-2,2,2-trichloroethane-d8, 1,1-Bis(p-hydroxyphenyl)-2,2,2-trichloroethane-d8, 2,2,2-Trichloro-1,1-bis(4-hydroxyphenyl)ethane-d8, α,α-Bis(4-hydroxyphenyl)-β,β,β-trichloroethane-d8
InChI formel InChI=1S/C14H11Cl3O2/c15-14(16,17)13(9-1-5-11(18)6-2-9)10-3-7-12(19)8-4-10/h1-8,13,18-19H/i1D,2D,3D,4D,5D,6D,7D,8D
SMIL OC1=C([2H])C([2H])=C(C(C(Cl)(Cl)Cl)C2=C([2H])C([2H])=C(O)C([2H])=C2[2H])C([2H])=C1[2H]
IUPAC navn 2,3,5,6-tetradeuterio-4-[2,2,2-trichloro-1-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)ethyl]phenol
Molekylvægt (g/mol) 325.64