Få mere at vide
2,2'-Bipyridine-5,5'-dicarboxylic Acid (6,6'-Dinicotinic Acid), Mikromol™
Artikelnummer. 30052560
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052560 100 mg
30050833 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052560 Leverandør Mikromol Leverandørnr. MM0553.06
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2,2'-Bipyridine-5,5'-dicarboxylic Acid (6,6'-Dinicotinic Acid)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,2'-Bipyridine-5,5'-dicarboxylic Acid (6,6'-Dinicotinic Acid)
CAS 1802-30-8
Analyt- eller komponentnavne 2,2'-Bipyridine-5,5'-dicarboxylic Acid (6,6'-Dinicotinic Acid)
Forsendelsestilstand Room Temperature
Molekylær formel C12 H8 N2 O4
Certificeringer/compliance ISO 17025
Synonym Nicotinic Acid Imp. B (EP), 6,6'-Dinicotinic Acid, 2,2'-Bipyridine-5,5'-dicarboxylic Acid, Nicotinic Acid Impurity B
InChI formel InChI=1S/C12H8N2O4/c15-11(16)7-1-3-9(13-5-7)10-4-2-8(6-14-10)12(17)18/h1-6H,(H,15,16)(H,17,18)
SMIL OC(=O)c1ccc(nc1)c2ccc(cn2)C(=O)O
IUPAC navn 6-(5-carboxypyridin-2-yl)pyridine-3-carboxylic acid
Molekylvægt (g/mol) 244.2
Mængde 100 mg
Formel vægt 244.0484 g/mol