Få mere at vide
2,2'-Binaphthyl-d14, TRC
Artikelnummer. 30344102
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30344102 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30344102 Leverandør Toronto Research Chemicals Leverandørnr. TRCB21119110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,2'-Binaphthyl-d14
CAS 210487-05-1
Molekylær formel C20 D14
Mængde 10 mg
Synonym 2,2'-Binaphthalene-1,1',3,3',4,4',5,5',6,6',7,7',8,8'-d14 (9CI), 2,2'-Binaphthalene-d14, 2,2'-Binaphthyl-D14, 2,2'-Dinaphthyl-d14, 2-(Naphthalen-2-yl-1,3,4,5,6,7,8-d7)naphthalene-1,3,4,5,6,7,8-d7, β,β'-Binaphthyl-d14
InChI formel InChI=1S/C20H14/c1-3-7-17-13-19(11-9-15(17)5-1)20-12-10-16-6-2-4-8-18(16)14-20/h1-14H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D,13D,14D
SMIL [2H]c1c([2H])c([2H])c2c([2H])c(c([2H])c([2H])c2c1[2H])c3c([2H])c([2H])c4c([2H])c([2H])c([2H])c([2H])c4c3[2H]
IUPAC navn 1,2,3,4,5,6,8-heptadeuterio-7-(1,3,4,5,6,7,8-heptadeuterionaphthalen-2-yl)naphthalene
Molekylvægt (g/mol) 268.4114
Formel vægt 268.1974