missing translation for 'onlineSavingsMsg'
Få mere at vide

2-(2-Benzylphenoxy)-N-methylethanamine Hydrochloride, Mikromol™

Artikelnummer. 30050311
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050311 25 mg
30051763 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30050311 Leverandør Mikromol Leverandørnr. MM0770.010025

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(2-Benzylphenoxy)-N-methylethanamine Hydrochloride
TRUSTED_SUSTAINABILITY

Especificaciones

Kemisk navn eller materiale Phenyltoloxamine Related Compound A
CAS 75859-72-2
Analyt- eller komponentnavne Phenyltoloxamine Related Compound A
Forsendelsestilstand Room Temperature
Molekylær formel C16 H19 N O . Cl H
Certificeringer/compliance ISO 17025
Synonym Ethanamine, N-methyl-2-[2-(phenylmethyl)phenoxy]-, hydrochloride (1:1), Ethanamine, N-methyl-2-[2-(phenylmethyl)phenoxy]-, hydrochloride (9CI), Ethylamine, N-methyl-2-(α-phenyl-o-tolyloxy)-, hydrochloride (5CI), Phenyltoloxamine Related Compound A as Hydrochloride
InChI formel InChI=1S/C16H19NO.ClH/c1-17-11-12-18-16-10-6-5-9-15(16)13-14-7-3-2-4-8-14;/h2-10,17H,11-13H2,1H3;1H
SMIL Cl.CNCCOc1ccccc1Cc2ccccc2
IUPAC navn 2-(2-benzylphenoxy)-N-methylethanamine;hydrochloride
Molekylvægt (g/mol) 277.79
Mængde 25 mg
Formel vægt 277.1233 g/mol
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.