missing translation for 'onlineSavingsMsg'
Learn More

2,2', Azino-bis-(3-ethylbenzthiazoline sulfonic acid), diammonium salt, For TLC analysis, MP Biomedicals™

Catalog Number p-4405764
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
30931-67-0
C18H24N6O6S4
548.67
MFCD00010404,MFCD00010404
OHDRQQURAXLVGJ-AXMZSLBLSA-N
ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate
91884754
diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate
[NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O
This item is not returnable. View return policy
30931-67-0
C18H24N6O6S4
548.67
MFCD00010404,MFCD00010404
OHDRQQURAXLVGJ-AXMZSLBLSA-N
ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate
91884754
diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate
[NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O
Color Green
Quantity 1 g
Formula Weight 548.7
Physical Form Powder
Chemical Name or Material 2, 2' Azino-bis-(3-ethylbenzthiazoline sulfonic acid), diammonium salt, For TLC analysis
compliance-icons
Product Identifier
  • 2,2'-Azino-bis(3-ethylbenzthiazoline-6-sulfonic Acid) Diammonium Salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'