Få mere at vide
2-(2-Aminoethyl)-3',6'-bis(diethylamino)spiro[isoindoline-1,9'-xanthen]-3-one (>80%), TRC
Artikelnummer. 30319179
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319179 100 mg
30315226 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30319179 Leverandør Toronto Research Chemicals Leverandørnr. TRCA622790100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(2-Aminoethyl)-3',6'-bis(diethylamino)spiro[isoindoline-1,9'-xanthen]-3-one
CAS 950846-89-6
Molekylær formel C30H36N4O2
Mængde 100 mg
Synonym [9H]xanthen]-3(2H)-one, Rhodamine B ethylenediamine
InChI formel InChI=1S/C30H36N4O2/c1-5-32(6-2)21-13-15-25-27(19-21)36-28-20-22(33(7-3)8-4)14-16-26(28)30(25)24-12-10-9-11-23(24)29(35)34(30)18-17-31/h9-16,19-20H,5-8,17-18,31H2,1-4H3
SMIL CCN(CC)c1ccc2c(Oc3cc(ccc3C24N(CCN)C(=O)c5ccccc45)N(CC)CC)c1
IUPAC navn 2-(2-aminoethyl)-3',6'-bis(diethylamino)spiro[isoindole-3,9'-xanthene]-1-one
Molekylvægt (g/mol) 484.63
Formel vægt 484.2838