Learn More
2-(2-Aminoethoxy)ethanol, Sigma-Aldrich™
Product Code. 15864411
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15864411 500 mL 500mL
15854411 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15864411 Supplier Sigma Aldrich Supplier No. 8184310500
 Restricted Item
Forventet forsendelse: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 929-06-6
Grade Research
Molecular Weight (g/mol) 105.14
Specifications
Melting Point -11°C
Density 1.06 g/cm3 at 20°C
Boiling Point 218°C to 224°C/1013 mbar
MDL Number MFCD00008181
Quantity 500 mL
Synonym 2-(2-Aminoethoxy)ethanol, Ethylene glycol mono(2-aminoethyl) ether, 2-(2-Aminoethoxy)ethanol