Få mere at vide
[2-[2-Amino-3-hydroxy-2-(hydroxymethyl)propoxy]-1-hydroxypropyl]phosphonic Acid, >90%, TRC
Artikelnummer. 30007774
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007774 5 mg
30006762 10 mg
30007410 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007774 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6115655MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1262243-11-7
Molekylær formel C7 H18 N O7 P
Molekylvægt (g/mol) 259.19
Synonym Fosfomycin EP-Impurity B
IUPAC navn [2-[2-amino-3-hydroxy-2-(hydroxymethyl)propoxy]-1-hydroxypropyl]phosphonic acid
SMIL CC(OCC(N)(CO)CO)C(O)P(=O)(O)O
Tekniske data
Kemisk navn eller materiale (2-[2-Amino-3-hydroxy-2-(hydroxymethyl)propoxy]-1-hydroxypropyl)phosphonic Acid
Analyt- eller komponentnavne (2-[2-Amino-3-hydroxy-2-(hydroxymethyl)propoxy]-1-hydroxypropyl)phosphonic Acid
Renhedsgrad noter HPLC
Forsendelsestilstand Dry Pack
InChI formel InChI=1S/C7H18NO7P/c1-5(6(11)16(12,13)14)15-4-7(8,2-9)3-10/h5-6,9-11H,2-4,8H2,1H3,(H2,12,13,14)
Løsningstype Neat
Industry Type Forensics
Mængde 5 mg
Formel vægt 259.0821
Procent renhed >95%