Learn More
2-(2-Amino-2-propyl)aniline, 95%, Thermo Scientific™
Catalog Number 15437597
Click to view available options
Quantity:
250 mg
1 g
229326-17-4
C9H14N2
150.23
MFCD08276909
MWUKUEYFFYQSQE-UHFFFAOYSA-N
2-2-aminopropan-2-yl aniline, 2-2-amino-2-propyl aniline, ambkkkkk207, 2-2-amino-2-propanyl aniline, 2-2-aminophenyl-2-propylamine, 2-amino-alpha,alpha-dimethylbenzylamine, benzenemethanamine, 2-amino-alpha,alpha-dimethyl-9ci
21719463
2-(2-aminopropan-2-yl)aniline
CC(C)(N)C1=CC=CC=C1N
229326-17-4
C9H14N2
150.23
MFCD08276909
MWUKUEYFFYQSQE-UHFFFAOYSA-N
2-2-aminopropan-2-yl aniline, 2-2-amino-2-propyl aniline, ambkkkkk207, 2-2-amino-2-propanyl aniline, 2-2-aminophenyl-2-propylamine, 2-amino-alpha,alpha-dimethylbenzylamine, benzenemethanamine, 2-amino-alpha,alpha-dimethyl-9ci
21719463
2-(2-aminopropan-2-yl)aniline
CC(C)(N)C1=CC=CC=C1N
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2810
Formula Weight 150.22
Percent Purity 95%
Chemical Name or Material 2-(2-Amino-2-propyl)aniline