Få mere at vide
2-[(2-Amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethylester N-Methyl-L-Valine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Aciclovir N-Methyl-L-valinate |
| CAS | 1346747-65-6 |
| Molekylær formel | C14 H22 N6 O4 |
| Mængde | 10 mg |
| Synonym | L-Valine, N-methyl-, 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethyl ester, 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-Methyl-L-valinate, Aciclovir N-Methyl-L-valinate, Valacyclovir USP Related Compound C |
| InChI formel | InChI=1S/C14H22N6O4/c1-8(2)9(16-3)13(22)24-5-4-23-7-20-6-17-10-11(20)18-14(15)19-12(10)21/h6,8-9,16H,4-5,7H2,1-3H3,(H3,15,18,19,21)/t9-/m0/s1 |
| SMIL | CN[C@@H](C(C)C)C(=O)OCCOCn1cnc2C(=O)NC(=Nc12)N |
| IUPAC navn | 2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]ethyl (2S)-3-methyl-2-(methylamino)butanoate |
| Molekylvægt (g/mol) | 338.36 |
| Formel vægt | 338.1703 |