Få mere at vide
2-[(2,6-Dichlorophenyl)amino]benzaldehyde, LoGiCal
Artikelnummer. 30459719
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459719 10 mg
1 options
Artikelnummer. 30459719 Leverandør LoGiCal Leverandørnr. LGCFOR0006.03
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[(2,6-Dichlorophenyl)amino]benzaldehyde
CAS 22121-58-0
Analyt- eller komponentnavne 2-[(2,6-Dichlorophenyl)amino]benzaldehyde
Forsendelsestilstand Room Temperature
Molekylær formel C13 H9 Cl2 N O
Certificeringer/compliance ISO 9001
Synonym Diclofenac Impurity B, Diclofenac Sodium Imp. B (EP), Diclofenac Imp. B (EP), 2-[(2,6-Dichlorophenyl)amino]benzaldehyde, Anthranilaldehyde, N-(2,6-dichlorophenyl)- (8CI), 2-(2,6-Dichloroanilino)benzaldehyde, o-(2,6-Dichloroanilino)benzaldehyde, Diclofenac Sodium Impurity B, Diclofenac Impurity B
InChI formel InChI=1S/C13H9Cl2NO/c14-10-5-3-6-11(15)13(10)16-12-7-2-1-4-9(12)8-17/h1-8,16H
Løsningstype Neat
SMIL Clc1cccc(Cl)c1Nc2ccccc2C=O
IUPAC navn 2-(2,6-dichloroanilino)benzaldehyde
Industry Type Clinical Testing
Molekylvægt (g/mol) 266.12
Mængde 10 mg
Formel vægt 265.0061 g/mol