missing translation for 'onlineSavingsMsg'
Få mere at vide

2,2',5'-Trichloroacetophenone, TRC

Artikelnummer. 30455844
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30455844 25 mg
30454715 250 mg
30455753 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30455844 Leverandør Toronto Research Chemicals Leverandørnr. TRCT89153025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 7396-79-4
Molekylær formel C8 H5 Cl3 O
Molekylvægt (g/mol) 223.48
Synonym 2-Chloro-1-(2,5-dichlorophenyl)ethanone, 2,2',5'-Trichloroacetophenone, 2,5,ω-Trichloroacetophenone, 2,5-Dichlorophenacyl chloride
IUPAC navn 2-chloro-1-(2,5-dichlorophenyl)ethanone
SMIL ClCC(=O)c1cc(Cl)ccc1Cl

Specifications

Kemisk navn eller materiale 2,2',5'-Trichloroacetophenone
Mængde 25 mg
InChI formel InChI=1S/C8H5Cl3O/c9-4-8(12)6-3-5(10)1-2-7(6)11/h1-3H,4H2
Formel vægt 221.9406
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.