Få mere at vide
2,2,4-Trimethylpentane-d18, CDN
Artikelnummer. 30317327
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30317327 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317327 Leverandør CDN Leverandørnr. CDND20101G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,2,4-Trimethylpentane (D18)
CAS 51685-57-5
Molekylær formel C8 D18
Mængde 1 g
Synonym Pentane-1,1,1,3,3,4,5,5,5-d9, 2,2,4-tri(methyl-d3)- (9 CI, ACI), 2,2,4-Tri(methyl-d3)pentane-1,1,1,3,3,4,5,5,5-d9 (ACI), Pentane-1,1,1,2,3,3,5,5,5-d9, tri(methyl-d3)- (7 CI), Pentane-1,1,1,2,3,3,5,5,6-d9, 2,4,4-tri(methyl-d3)- (6 CI), Isooctane-d18, Perdeuterated isooctane, 2,2,4-Trimethylpentane-d18, 2,2,4-Trimethylpentane-D18
InChI formel InChI=1 S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3/i1D3,2D3,3D3,4D3,5D3,6D2,7 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,2,3,3,5,5,5-nonadeuterio-2,4,4-tris(trideuteriomethyl)pentane
Molekylvægt (g/mol) 132.3394
Formel vægt 132.2538 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity