missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4-Trimethylpentane CHROMASOLV™, for HPLC, ≥99%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
100 mL
1 L
2 L
2.5 L
4 x 2 L
4 x 2.5 L
Unit Size:
1 L
100mL
2 L
2.50 L
Pack of 4
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency

Specifications
Specifications
Melting Point | -107°C |
Density | 0.692g/mL at 25°C |
Boiling Point | 98°C to 99°C |
Quantity | 2 L |
Refractive Index | n20/D 1.391 |
Linear Formula | (CH3)2CHCH2C(CH3)3 |
Vapor Pressure | 41mmHg (21 °C) |
Beilstein | 1696876 |
Formula Weight | 114.23g/mol |
Percent Purity | ≥99% |
Show More |
Safety and Handling




Product Identifier
- 2,2,4-Trimethylpentane
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,2,4-Trimethylpentane CHROMASOLV™, for HPLC, ≥99%, Honeywell Riedel-de Haën™ >
Spot an opportunity for improvement?Share a Content Correction