Få mere at vide
2-[2-[4-[(R)-(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-acetamide, TRC
Artikelnummer. 30368115
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30368115 10 mg -
30366900 25 mg -
30353280 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30368115 Leverandør Toronto Research Chemicals Leverandørnr. TRCC59649510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 909779-33-5
Molekylær formel C21 H26 Cl N3 O2
Molekylvægt (g/mol) 387.9
Synonym Levocetirizine Amide, (R)-2-(2-{4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl}ethoxy)acetamide, Acetamide, 2-[2-[4-[(R)-(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-
IUPAC navn 2-[2-[4-[(R)-(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetamide
SMIL NC(=O)COCCN1CCN(CC1)[C@H](c2ccccc2)c3ccc(Cl)cc3
Tekniske data
Kemisk navn eller materiale Levocetirizine Amide
Mængde 10 mg
InChI formel InChI=1S/C21H26ClN3O2/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)25-12-10-24(11-13-25)14-15-27-16-20(23)26/h1-9,21H,10-16H2,(H2,23,26)/t21-/m1/s1
Formel vægt 387.1714