Få mere at vide
2-[2-(4-Fluorophenyl)-2-oxo-1-phenylethyl]-4-methyl-3-oxo-N-phenylpentanamide (4-Fluoro-alpha-(2-methyl-1-oxopropyl)-γ-oxo-N,beta-diphenylbenzenebutanamide), Mikromol™
Artikelnummer. 30049111
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049111 25 mg
17979954 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049111 Leverandør Mikromol Leverandørnr. MM0400.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[2-(4-Fluorophenyl)-2-oxo-1-phenylethyl]-4-methyl-3-oxo-N-phenylpentanamide (4-Fluoro-alpha-(2-methyl-1-oxopropyl)-γ-oxo-N,beta-diphenylbenzenebutanamide)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Fluoro-alpha-(2-methyl-1-oxopropyl)-gamma-oxo-N,beta-diphenylbenzene butanamide
CAS 125971-96-2
Analyt- eller komponentnavne 4-Fluoro-alpha-(2-methyl-1-oxopropyl)-gamma-oxo-N,beta-diphenylbenzene butanamide
Forsendelsestilstand Room Temperature
Molekylær formel C26 H24 F N O3
Certificeringer/compliance ISO 17025
Synonym 2-[2-(4-Fluorophenyl)-2-oxo-1-phenylethyl]-4-methyl-3-oxopentanoic acid phenylamide, 4-Fluoro-alpha-(2-methyl-1-oxopropyl)-gamma-oxo-N,beta-diphenylbenzenebutanamide
InChI formel InChI=1S/C26H24FNO3/c1-17(2)24(29)23(26(31)28-21-11-7-4-8-12-21)22(18-9-5-3-6-10-18)25(30)19-13-15-20(27)16-14-19/h3-17,22-23H,1-2H3,(H,28,31)
SMIL CC(C)C(=O)C(C(C(=O)c1ccc(F)cc1)c2ccccc2)C(=O)Nc3ccccc3
IUPAC navn 2-[2-(4-fluorophenyl)-2-oxo-1-phenylethyl]-4-methyl-3-oxo-N-phenylpentanamide
Molekylvægt (g/mol) 417.47
Mængde 25 mg
Formel vægt 417.174 g/mol