missing translation for 'onlineSavingsMsg'
Få mere at vide

2-(2,4-Difluorophenyl)propan-2-ol, TRC

Artikelnummer. 30385890
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30385890 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30385890 Leverandør Toronto Research Chemicals Leverandørnr. TRCD445725100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-(2,4-Difluorophenyl)propan-2-ol
CAS 51788-81-9
Molekylær formel C9H10F2O
Mængde 100 mg
Synonym 2,4-Difluoro-alpha,alpha-dimethyl-benzenemethanol
InChI formel InChI=1S/C9H10F2O/c1-9(2,12)7-4-3-6(10)5-8(7)11/h3-5,12H,1-2H3
SMIL CC(C)(O)c1ccc(F)cc1F
Molekylvægt (g/mol) 172.17
Formel vægt 172.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.