missing translation for 'onlineSavingsMsg'
Få mere at vide

2,2,4,6,7-Pentamethyl-2,3-dihydrobenzofuran-5-sulfonyl Chloride (>80%), TRC

Artikelnummer. 30347348
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30347348 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30347348 Leverandør Toronto Research Chemicals Leverandørnr. TRCB510560500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2,2,4,6,7-Pentamethyl-2,3-dihydrobenzofuran-5-sulfonyl Chloride
CAS 154445-78-0
Molekylær formel C13 H17 Cl O3 S
Mængde 500 mg
InChI formel InChI=1S/C13H17ClO3S/c1-7-8(2)12(18(14,15)16)9(3)10-6-13(4,5)17-11(7)10/h6H2,1-5H3
SMIL Cc1c(C)c(c(C)c2CC(C)(C)Oc12)S(=O)(=O)Cl
IUPAC navn 2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-sulfonyl chloride
Molekylvægt (g/mol) 288.79
Formel vægt 288.059
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.