Få mere at vide
2,2',4,5,5'-Pentachlorobiphenyl-3',4',6'-d3, TRC
Artikelnummer. 30413663
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30413663 10 mg
30409653 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30413663 Leverandør Toronto Research Chemicals Leverandørnr. TRCP22041310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1219794-68-9
Molekylær formel C12 D3 H2 Cl5
Molekylvægt (g/mol) 329.452
Synonym 2,2',4,5,5'-Pentachlorobiphenyl-3',4',6'-D3, PCB 101 D3, 2,2',4,5,5'-PentaCB-3',4',6'-d3, PCB No. 101-d3
IUPAC navn 1,4-dichloro-2,3,5-trideuterio-6-(2,4,5-trichlorophenyl)benzene
SMIL [2H]c1c([2H])c(Cl)c(c([2H])c1Cl)c2cc(Cl)c(Cl)cc2Cl
Tekniske data
Kemisk navn eller materiale 2,2',4,5,5'-Pentachlorobiphenyl-3',4',6'-d3
Mængde 10 mg
InChI formel InChI=1S/C12H5Cl5/c13-6-1-2-9(14)7(3-6)8-4-11(16)12(17)5-10(8)15/h1-5H/i1D,2D,3D
Formel vægt 326.902