Learn More
2,2',4,4'-Tetrahydroxybenzophenone, 98+%, Thermo Scientific Chemicals
Catalog Number p-7080630
Click to view available options
Quantity:
25 g
100 g
131-55-5
C13H10O5
246.218
MFCD00002278
WXNRYSGJLQFHBR-UHFFFAOYSA-N
2,2',4,4'-tetrahydroxybenzophenone, bis 2,4-dihydroxyphenyl methanone, benzophenone-2, uvinol d-50, uvinul d-50, methanone, bis 2,4-dihydroxyphenyl, 2,4,2',4'-tetrahydroxybenzophenone, unii-prr8k3h9vn, 2,2',4,4'-tetrehydroxybenzophenone, prr8k3h9vn
8571
bis(2,4-dihydroxyphenyl)methanone
C1=CC(=C(C=C1O)O)C(=O)C2=C(C=C(C=C2)O)O
131-55-5
C13H10O5
246.218
MFCD00002278
WXNRYSGJLQFHBR-UHFFFAOYSA-N
2,2',4,4'-tetrahydroxybenzophenone, bis 2,4-dihydroxyphenyl methanone, benzophenone-2, uvinol d-50, uvinul d-50, methanone, bis 2,4-dihydroxyphenyl, 2,4,2',4'-tetrahydroxybenzophenone, unii-prr8k3h9vn, 2,2',4,4'-tetrehydroxybenzophenone, prr8k3h9vn
8571
bis(2,4-dihydroxyphenyl)methanone
C1=CC(=C(C=C1O)O)C(=O)C2=C(C=C(C=C2)O)O
2,2',4,4'-Tetrahydroxybenzophenone is used as the raw material of ultraviolet absorbers in cosmetics, plastics, resins and rubber. It is used as pharmaceutical intermediates and light-sensitive materials. It is also used as pharmaceutical adjuvant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2′,4,4′-Tetrahydroxybenzophenone is used as the raw material of ultraviolet absorbers in cosmetics, plastics, resins and rubber. It is used as pharmaceutical intermediates and light-sensitive materials. It is also used as pharmaceutical adjuvant.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 198°C to 203°C
Density 1.216
Quantity 25 g
Beilstein 1914746
Solubility Information Slightly soluble in water.
Formula Weight 246.22
Percent Purity ≥98%
Chemical Name or Material 2,2',4,4'-Tetrahydroxybenzophenone