Få mere at vide
2,2,4,4,6,6-Hexakis-phenyl-1,3,5,2,4,6-trioxatrisilinane, TRC
Artikelnummer. 30180410
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180410 250 mg
30179904 100 mg
30180174 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180410 Leverandør Toronto Research Chemicals Leverandørnr. TRCH291018250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 512-63-0
Molekylær formel C36 H30 O3 Si3
Molekylvægt (g/mol) 594.878
Synonym hexaphenyl-Cyclotrisiloxane, 2,2,4,4,6,6-Hexaphenylcyclotrisiloxane, Diphenylsiloxane cyclic trimer, Hexaphenylcyclotrisiloxane, NSC 127080
IUPAC navn 2,2,4,4,6,6-hexakis-phenyl-1,3,5,2,4,6-trioxatrisilinane
SMIL O1[Si](O[Si](O[Si]1(c2ccccc2)c3ccccc3)(c4ccccc4)c5ccccc5)(c6ccccc6)c7ccccc7
Tekniske data
Kemisk navn eller materiale 2,2,4,4,6,6-Hexakis-phenyl-1,3,5,2,4,6-trioxatrisilinane
Mængde 250 mg
InChI formel InChI=1S/C36H30O3Si3/c1-7-19-31(20-8-1)40(32-21-9-2-10-22-32)37-41(33-23-11-3-12-24-33,34-25-13-4-14-26-34)39-42(38-40,35-27-15-5-16-28-35)36-29-17-6-18-30-36/h1-30H
Formel vægt 594.15