missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,3,3,4,4,4-Heptafluorobutyl acrylate, 97%, stab. with 100 ppm 4-methoxyphenol, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2,2,3,3,4,4,4-Heptafluorobutyl acrylate is a useful fluorinated acrylate for proteomics research. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2,3,3,4,4,4-Heptafluorobutyl acrylate is a useful fluorinated acrylate for proteomics research. It is also used as pharmaceutical intermediate.
Solubility
Not miscible in water.
Notes
Light sensitive. Incompatible with oxidizing agents, heat, ultraviolet radiation and free radical initiators.
2,2,3,3,4,4,4-Heptafluorobutyl acrylate is a useful fluorinated acrylate for proteomics research. It is also used as pharmaceutical intermediate.
Solubility
Not miscible in water.
Notes
Light sensitive. Incompatible with oxidizing agents, heat, ultraviolet radiation and free radical initiators.
Specifications
Density | 1.418 |
Boiling Point | 57°C to 58°C (30 mmHg) |
Flash Point | 31°C (87°F) |
Odor | Pungent |
Refractive Index | 1.331 |
Quantity | 5 g |
UN Number | UN3272 |
Beilstein | 1792520 |
Solubility Information | Not miscible in water. |
Formula Weight | 254.11 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'